Key Documents
SYNA551420
Synthware distillation distribution adapter
joint: ST/NS 14/20
Synonym(s):
Synthware A551420, cow distillation adapter
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A551420
General description
Rotating cow receiver allows four flasks to be progressively filled.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)