Skip to Content
Merck
All Photos(1)

Key Documents

SLW3730/06

Pyrex® Filter funnel with sintered glass disc

20 mL, bowl I.D. 65 mm, porosity grade 3

Synonym(s):

Pyrex, SciLabware 3730/06

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.43

material

borosilicate glass

manufacturer/tradename

3730/06
SciLabware

bowl I.D.

65 mm

capacity

20 mL
20 mL

stem O.D. × L

8 mm × 75 mm

pore size

grade 3 porosity

Looking for similar products? Visit Product Comparison Guide

General description

  • Sintered disc manufactured from Pyrex borosilicate glass
  • Available in a choice of porosities

For the filtration of solutions:
  • No requirement for filter paper
  • Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service