Key Documents
SYNC144250
Synthware Graham condenser
joint: ST/NS 24/40, hose size 10 mm
Synonym(s):
Synthware C144250, distillation condenser, Inland Revenue condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C144250
hose size
10 mm
Related Categories
General description
Synthware Graham condenser features a 250 mm jacketed length for superior cooling efficiency. With standardized 24/40 upper and lower joints, it easily fits various laboratory glassware. Made from durable borosilicate glass.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)