Skip to Content
Merck
All Photos(1)

Key Documents

QBD10554

Sigma-Aldrich

MAL-dPEG®24-TFP ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H108F4N2O29
Molecular Weight:
1445.53
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reactivity: thiol reactive
reagent type: cross-linking reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(OC1=C(C(F)=CC(F)=C1F)F)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(CCN2C(C=CC2=O)=O)=O

Features and Benefits

MAL-dPEG®24-TFP esters is a heterobifunctional, linear crosslinking reagent containing a maleimide group on one end of the single molecular weight dPEG® spacer arm and an amine-reactive active ester on the other end. The maleimide group reacts readily and specifically with free sulfhydryl groups in aqueous media at pH 6.5-7.5. 2,3,5,6-tetrafluorophenyl (TFP) ester is used widely in reactions with primary amines. The TFP-amine reactions can be carried out in aqueous or organic solvent. The dPEG® spacer is hydrophilic and non-immunogenic. Because it is a single molecular weight product, it gives the user precise spatial control. The dPEG® spacer also improves water solubility, decreases immunogenicity, and increases the hydrodynamic volume of the conjugate. The increased hydrodynamic volume results in longer serum half-life in vivo.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service