Skip to Content
Merck
All Photos(1)

Key Documents

P4607

Sigma-Aldrich

1,4-Pentadiene

99%

Synonym(s):

Allylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2CH=CH
CAS Number:
Molecular Weight:
68.12
Beilstein:
1696934
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

11.91 psi ( 20 °C)

Quality Level

Assay

99%

refractive index

n20/D 1.389 (lit.)

bp

26 °C (lit.)

mp

−148 °C (lit.)

density

0.659 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CCC=C

InChI

1S/C5H8/c1-3-5-4-2/h3-4H,1-2,5H2

InChI key

QYZLKGVUSQXAMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Pentadiene can be used as a precursor in the synthesis of (+)-isolysergol, phosphorinanes and lithium pentadienyl complexes.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Light-Driven, Zirconium-Catalyzed Hydrophosphination with Primary Phosphines.
Bange C A, et al.
ACS Catalysis (2018)
A H Neerincx et al.
Journal of breath research, 10(4), 046014-046014 (2016-12-03)
Staphylococcus aureus (S. aureus) is a common bacterium infecting children with cystic fibrosis (CF). Since current detection methods are difficult to perform in children, there is need for an alternative. This proof of concept study investigates whether breath profiles can
Fethi Khaled et al.
The journal of physical chemistry. A, 123(11), 2261-2271 (2019-02-16)
Hydroxyl radicals and olefins are quite important from a combustion and an atmospheric chemistry standpoint. Large amounts of olefinic compounds are emitted into the earth's atmosphere from both biogenic and anthropogenic sources. Olefins make a significant share in transportation fuels
Structural Influences in Lithium Pentadienylsilane Complexes.
Day B M, et al.
Organometallics, 32(15), 4448-4451 (2013)
Jason A Deck et al.
Organic letters, 12(11), 2610-2613 (2010-05-14)
The first enantioselective synthesis of (+)-isolysergol was completed in 12 steps from commercially available materials by a novel approach that features a late stage microwave-mediated, diastereomeric ring-closing metathesis catalyzed by a chiral molybdenum catalyst to simultaneously form the D ring

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service