Skip to Content
Merck
All Photos(1)

Key Documents

333646

Sigma-Aldrich

1,13-Tetradecadiene

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)10CH=CH2
CAS Number:
Molecular Weight:
194.36
Beilstein:
1741459
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

liquid

refractive index

n20/D 1.443 (lit.)

bp

131 °C/17 mmHg (lit.)

density

0.849 g/mL at 25 °C (lit.)

functional group

allyl

SMILES string

C=CCCCCCCCCCCC=C

InChI

1S/C14H26/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3-4H,1-2,5-14H2

InChI key

XMRSTLBCBDIKFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,13-Tetradecadiene is an α-diolefin. Polymerization of 1,13-tetradecadiene using aluminum triisobutyl-titanium tetrachloride was reported.

Application

1,13-Tetradecadiene was grafted to hydrogenated B-doped silicon (100) surfaces. It was used in the synthesis of naturally occurring, biologically active pyridine alkaloids theonelladins C and D, niphatesine C and xestamine D.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymerization of Higher a-Diolefins with Metal Alkyl Coordination Catalysts1.
Marvel CS and Garrison Jr WE.
Journal of the American Chemical Society, 81(17), 4737-4744 (1959)
Monolayers of simple organic molecules on silicon studied by surface tools.
Scandurra A, et al.
Surface and Interface Analysis : SIA, 34(1), 777-777 (2002)
Synthesis of pyridine alkaloids via Pd-catalyzed coupling of 3-iodopyridine, 1, ?-dienes and nitrogen nucleophiles.
Larock RC and Wang Y.
Tetrahedron Letters, 43(1), 21-23 (2002)
Kotohiro Nomura et al.
Polymers, 12(1) (2019-12-22)
Copolymerizations of 1-decene (DC) with 1,9-decadiene (DCD), 1-dodecene (DD) with 1,11-dodecadiene (DDD), and 1-tetradecene (TD) with 1,13-tetradecadiene (TDD), using Cp*TiMe2(O-2,6-iPr2C6H3) (1)-[Ph3C][B(C6F5)4] (borate) catalyst in the presence of AliBu3/Al(n-C8H17)3 proceeded in a quasi-living manner in n-hexane at -30 to -50 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service