P30802
2-Phenyl-2-propanol
97%
Synonym(s):
α,α-Dimethylbenzyl alcohol, Dimethyl phenyl carbinol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2C(OH)C6H5
CAS Number:
Molecular Weight:
136.19
Beilstein:
1905012
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
powder
refractive index
n20/D 1.5196 (lit.)
bp
202 °C (lit.)
mp
28-32 °C (lit.)
density
0.973 g/mL at 25 °C (lit.)
SMILES string
CC(C)(O)c1ccccc1
InChI
1S/C9H12O/c1-9(2,10)8-6-4-3-5-7-8/h3-7,10H,1-2H3
InChI key
BDCFWIDZNLCTMF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
[Photometric determination of dimethylphenylcarbinol in water].
Z A Shcherbakova
Gigiena i sanitariia, (1)(1), 71-72 (1982-01-01)
[Information from the Soviet Toxicology Center].
Gigiena truda i professional'nye zabolevaniia, (8)(8), 53-56 (1982-08-01)
J Scognamiglio et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S130-S133 (2011-10-29)
A toxicologic and dermatologic review of 2-phenyl-2-propanol when used as a fragrance ingredient is presented. 2-Phenyl-2-propanol is a member of the fragrance structural group Aryl Alkyl Alcohols and is a tertiary alcohol. The AAAs are a structurally diverse class of
[Toxicological evaluation of the plasticizer PB-I].
Iu I Stepkin et al.
Gigiena i sanitariia, (4)(4), 50-52 (2000-09-07)
Jiafeng Geng et al.
Journal of inorganic biochemistry, 167, 60-67 (2016-12-03)
The diheme enzyme MauG utilizes H
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service