Skip to Content
Merck
All Photos(2)

Key Documents

M33476

Sigma-Aldrich

2-Methylbutyraldehyde

95%

Synonym(s):

α-Methyl-n-butanal, α-Methylbutanal, α-Methylbutyric aldehyde, (±)-2-Methylbutanal, 2-Ethylpropanal, 2-Formylbutane, 2-Methylbutanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CHO
CAS Number:
Molecular Weight:
86.13
Beilstein:
1633540
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.3919 (lit.)

bp

90-92 °C (lit.)

density

0.806 g/mL at 20 °C
0.804 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)C(C)CC

InChI

1S/C5H10O/c1-3-5(2)4-6/h4-5H,3H2,1-2H3

InChI key

BYGQBDHUGHBGMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methylbutyraldehyde can be used as a reactant to synthesize:
  • 2-amino-3-cyanopyridine derivatives via multicomponent condensation with various acetophenones and malononitrile in the presence of ammonium acetate.
  • N,N-Diethyl-2-methyl-1-butanamine by reacting with diethylamine via Rh-catalyzed reductive amination reaction in the presence of molecular hydrogen.
  • α-(2-Methylbutylidene)benzeneacetonitrile by condensation reaction with benzyl cyanide using tricyclic strong nonionic Lewis bases.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

23.0 °F

Flash Point(C)

-5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient synthesis of 3-cyano-6-(2-hydroxyphenyl) pyridines by multi-component condensations on beads
Shintani T, et al.
Tetrahedron Letters, 44(35), 6567-6569 (2003)
Direct synthesis of α ,β-unsaturated nitriles catalyzed by nonionic superbases
D'Sa Bosco A, et al.
The Journal of Organic Chemistry, 63(12), 3961-3967 (1998)
Rhodium-Catalysed Reductive Amination for the Synthesis of Tertiary Amines
Bianga J, et al.
Advanced Synthesis & Catalysis, 362(20), 4415-4424 (2020)
Rhodium-Catalysed Reductive Amination for the Synthesis of Tertiary Amines
Bianga J, et al.
advanced synthesis and catalysis, 362(20), 4415-4424 (2020)
Yunzi Feng et al.
Food chemistry, 187, 44-52 (2015-05-16)
Aroma extract dilution analysis (AEDA) is widely used for the screening of aroma-active compounds in gas chromatography-olfactometry (GC-O). In this study, three aroma dilution methods, (I) using different test sample volumes, (II) diluting samples, and (III) adjusting the GC injector

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service