HAM204001
Hamilton® chromatography syringe
for ThermoFinnigan GC Autosampler, model, 701 N, cemented needle, volume × L 10 μL × 50 mm, needle gauge 26s
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
Recommended Products
material
Microliter syringe
grade
for chromatography
feature
autoclavable: no
model 701 N, cemented needle
manufacturer/tradename
Hamilton
parameter
20-50 °C temp. range (68-122 °F)
2000 psig pressure
needle I.D. × O.D.
0.13 mm × 0.483 mm
needle gauge
26s
volume × L
10 μL × 50 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
S-Line syringes deliver exceptional performance for everyday use in HPLC autosampler injections at a great value. The C-Line syringe minimizes the potential for bent needles with a keyed flange design. X-Type syringes are designed for sensitive LC samples (i.e. proteins, peptides, phospholipids, smaller amines, etc.) with a specially coated needle and inner syringe barrel to minimize sample adsorption.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service