Skip to Content
Merck
All Photos(2)

Key Documents

H9750

Sigma-Aldrich

Holmium(III) oxide

≥99.9% (rare earth content, expressed as Ho2O3)

Synonym(s):

Rare earth

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ho2O3
CAS Number:
Molecular Weight:
377.86
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.9% (rare earth content, expressed as Ho2O3)

form

powder

reaction suitability

reagent type: catalyst
core: holmium

mp

2330 °C (lit.)

SMILES string

O=[Ho]O[Ho]=O

InChI

1S/2Ho.3O

InChI key

JYTUFVYWTIKZGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Holmium(III) oxide is a white crystalline powder and is a rare-earth metal oxide. Holmium(III) oxide is a highly stable and resistant to chemical attack. It has a high melting point of 2,380 °C and a high density of 8.8 g/cm3. Holmium(III) oxide is used as a raw material in the production of holmium and holmium alloys, as a magneto strictive material in sensors, and a catalyst in the production of synthetic fuels among other applications.

Application

Holmium(III)oxide can be used as:
  • A starting material to prepare Ho2O3-PVAfilm for generating stable Q-switching pulses in a thulium-doped fiber laser(TDFL) cavity.
  • A catalyst for oxidative coupling of methane.
It can also be used as a dopant in the preparation of:
  • Cadmium lead borate glasses.
  • Diamond/copper composites to eliminate interfacial defects through the spark plasma sintering (SPS) method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service