Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Holmium(III) oxide
Synonym(s):
Rare earth, Holmium(III) oxide
Linear Formula:
Ho
2
O
3
CAS Number:
12055-62-8
Molecular Weight:
377.86
EC Number:
235-015-3
Products
Brand
Sigma-Aldrich (4)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
H9750
Holmium(III) oxide,
≥99.9% (rare earth content, expressed as Ho
2
O
3
)
Expand
View Pricing
229679
Holmium(III) oxide,
powder, 99.999% trace metals basis
Expand
View Pricing
1.12469
Holmium(III) oxide,
LAB
Expand
View Pricing
637327
Holmium(III) oxide,
nanopowder, <100 nm avg. part. size (DLS), ≥99.9% trace metals basis
Expand
View Pricing