Skip to Content
Merck
All Photos(1)

Key Documents

GF90913855

Alumina/Silica/Boria

fiber, tex number 110, Al2O3 70%/SiO2 28%/B2O3 2%, length 20 m, filament diameter 0.011mm

Synonym(s):

AJ625710

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11181511
NACRES:
NA.23

form

fibers

manufacturer/tradename

Goodfellow 909-138-55

L

20 m

diam.

0.011 mm , filament diameter

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service