Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Alumina/Silica/Boria
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
GF97869964
Alumina/Silica/Boria,
fabric, weight.m-2 -400g, Al2O3 62%/SiO2 24%/B2O3 14%, weight 400 g, length 2 m, coil width 25.4 mm
Expand
View Pricing
GF33746081
Alumina/Silica/Boria,
fabric, Al2O3 62%/SiO2 24%/B2O3 14%, weight.m-2 -400g, length 5 m, coil width 25.4 mm
Expand
View Pricing
GF47218530
Alumina/Silica/Boria,
fiber, Al2O3 62%/SiO2 24%/B2O3 14%, tex number 680, length 10 m, number of filaments
Expand
View Pricing
GF49256332
Alumina/Silica/Boria,
fiber, Al2O3 70%/SiO2 28%/B2O3 2%, tex number 110, length 1 m, filament diameter 0.011mm
Expand
View Pricing
GF71127312
Alumina/Silica/Boria,
fiber, Al2O3 70%/SiO2 28%/B2O3 2%, tex number 110, length 10 m, filament diameter 0.011mm
Expand
View Pricing
GF15982959
Alumina/Silica/Boria,
fabric, Al2O3 62%/SiO2 24%/B2O3 14%, weight.m-2 -400g, length 1 m, coil width 25.4 mm
Expand
View Pricing
GF45568974
Alumina/Silica/Boria,
fiber, Al2O3 62%/SiO2 24%/B2O3 14%, tex number 100, length 5 m, filament diameter 0.011mm
Expand
View Pricing
GF96121661
Alumina/Silica/Boria,
fiber, Al2O3 62%/SiO2 24%/B2O3 14%, tex number 100, length 10 m, filament diameter 0.011mm
Expand
View Pricing
GF02316462
Alumina/Silica/Boria,
fiber, Al2O3 62%/SiO2 24%/B2O3 14%, tex number 680, length 20 m, number of filaments
Expand
View Pricing
GF29017380
Alumina/Silica/Boria,
fiber, Al2O3 70%/SiO2 28%/B2O3 2%, tex number 110, length 100 m, filament diameter 0.011mm
Expand
View Pricing
GF89483956
Alumina/Silica/Boria,
fiber, tex number 680, Al2O3 62%/SiO2 24%/B2O3 14%, length 1 m, number of filaments
Expand
View Pricing
GF90913855
Alumina/Silica/Boria,
fiber, tex number 110, Al2O3 70%/SiO2 28%/B2O3 2%, length 20 m, filament diameter 0.011mm
Expand
View Pricing
GF15208657
Alumina/Silica/Boria,
fiber, Al2O3 62%/SiO2 24%/B2O3 14%, tex number 100, length 20 m, filament diameter 0.011mm
Expand
View Pricing
GF30169010
Alumina/Silica/Boria,
fiber, Al2O3 70%/SiO2 28%/B2O3 2%, tex number 110, length 5 m, filament diameter 0.011mm
Expand
View Pricing
GF64232131
Alumina/Silica/Boria,
fiber, Al2O3 70%/SiO2 28%/B2O3 2%, tex number 110, length 50 m, filament diameter 0.011mm
Expand
View Pricing
GF86764551
Alumina/Silica/Boria,
fiber, Al2O3 62%/SiO2 24%/B2O3 14%, tex number 100, filament diameter 0.011mm, length 2 m
Expand
View Pricing
GF86777746
Alumina/Silica/Boria,
fiber, tex number 680, Al2O3 62%/SiO2 24%/B2O3 14%, number of filaments, length 5 m
Expand
View Pricing
GF23894118
Alumina/Silica/Boria,
fiber, Al2O3 62%/SiO2 24%/B2O3 14%, tex number 100, length 1 m, filament diameter 0.011mm
Expand
View Pricing
GF85494224
Alumina/Silica/Boria,
fiber, tex number 680, Al2O3 62%/SiO2 24%/B2O3 14%, length 2 m, number of filaments
Expand
View Pricing
GF57041355
Alumina/Silica/Boria,
fiber, Al2O3 70%/SiO2 28%/B2O3 2%, tex number 110, length 2 m, filament diameter 0.011mm
Expand
View Pricing