GF58057566
Polycarbonate - 30% Carbon Fiber Reinforced
(PC - 30% CFR), granule, 3 mm nominal granule size, weight 1000 g, black
Synonym(s):
CT356310
About This Item
Recommended Products
form
granular
manufacturer/tradename
Goodfellow 580-575-66
weight
1000 g
color
black
nominal granule size
3 mm
Related Categories
1 of 4
This Item | SAB2108308 | SAB2100822 | SAB4300358 |
---|---|---|---|
human | bovine, rat, mouse, rabbit, horse, guinea pig, human, dog | dog, human, rabbit, guinea pig, bovine | human |
polyclonal | polyclonal | polyclonal | polyclonal |
affinity isolated antibody | affinity isolated antibody | affinity isolated antibody | affinity isolated antibody |
rabbit | rabbit | rabbit | rabbit |
wet ice | wet ice | wet ice | wet ice |
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service