Skip to Content
Merck
All Photos(2)

Key Documents

SAB2100822

Sigma-Aldrich

Anti-FLI1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-EWSR2, Anti-Friend leukemia virus integration 1, Anti-SIC-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

dog, human, rabbit, guinea pig, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FLI1(2313)

Related Categories

General description

Fli-1 proto-oncogene, ETS transcription factor (FLI1) is encoded by the gene mapped to human chromosome 11. The encoded protein belongs to the ETS family of transcription factors. FLI1 consists of ETS DNA-binding domain at its C-terminal end and is mainly expressed in hematopoietic and vascular endothelial cells.

Immunogen

Synthetic peptide directed towards the middle region of human FLI1

Application

Anti-FLI1 antibody produced in rabbit has been used in chromatin immunoprecipitation.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.Chromatin immunoprecipitation (1 paper)
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)

Biochem/physiol Actions

Fli-1 proto-oncogene, ETS transcription factor (FLI1) acts as a sequence-specific transcriptional activator. It might be implicated in the development of both the haematopoietic and vascular systems. In addition, it also plays a vital role in megakaryopoiesis and platelet function. FLI1 functions as a regulator of essential midkine (MK) genes. Mutations in FLI1 is associated with the pathogenesis of Paris-Trousseau syndrome and congenital thrombocytopenia.

Sequence

Synthetic peptide located within the following region: FDFHGIAQALQPHPTESSMYKYPSDISYMPSYHAHQQKVNFVPPHPSSMP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EWS, but not EWS-FLI-1, is associated with both TFIID and RNA polymerase II: interactions between two members of the TET family, EWS and hTAFII68, and subunits of TFIID and RNA polymerase II complexes.
Bertolotti A
Molecular and Cellular Biology, 18(3), 1489-1497 (1998)
The ETS-domain transcription factor family.
Sharrocks AD
Nature Reviews in Molecular and Cell Biology, 2(11), 827-837 (2001)
Genotoxic stress inhibits Ewing sarcoma cell growth by modulating alternative pre-mRNA processing of the RNA helicase DHX9.
Fidaleo M
Oncotarget, 6(31), 31740-31757 (2015)
Gianfranco Matrone et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(31) (2021-08-01)
A network of molecular factors drives the development, differentiation, and maintenance of endothelial cells. Friend leukemia integration 1 transcription factor (FLI1) is a bona fide marker of endothelial cells during early development. In zebrafish Tg( f li1:EGFP) y1 , we
Macrothrombocytopenia and dense granule deficiency associated with FLI1 variants: ultrastructural and pathogenic features.
Saultier P
Haematologica, 102(6), 1006-1016 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service