GF08235457
Indium
foil, 50x50mm, thickness 2.0mm, as rolled, 99.999%
Synonym(s):
Indium, IN000300
About This Item
vapor pressure
<0.01 mmHg ( 25 °C)
Assay
99.999%
form
foil
manufacturer/tradename
Goodfellow 082-354-57
resistivity
8.37 μΩ-cm
L × W × thickness
50 mm × 50 mm × 2.0 mm
mp
156.6 °C (lit.)
density
7.3 g/mL at 25 °C (lit.)
SMILES string
[In]
InChI
1S/In
InChI key
APFVFJFRJDLVQX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service