DWKW985850
WHEATON® CryoELITE® cryovial with Loctagon™ Vial Skirt and label patch
round bottom polypropylene, capacity (2 mL), external thread, non-sterile
Synonym(s):
W985850
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121704
Recommended Products
material
round bottom polypropylene
sterility
non-sterile
packaging
pkg of 1000 ea
manufacturer/tradename
W985850
technique(s)
cryopreservation: suitable
capacity
(2 mL)
Looking for similar products? Visit Product Comparison Guide
General description
Used in biobanking and forensics for sample collection and cyrogenic storage.
Features and Benefits
- Unrivaled cap seal exceeds DOT and IATA regulations ensuring ultimate protection of samples during transportation and demanding freeze-thaw handling
- Made from low binding, cryogenic grade virgin polypropylene
- Lot certified RNase/DNase and Endotoxin Free providing assurance of product integrity
- Free standing vials feature a conical well design, which allows for precise sample retrieval from both automated and manual systems
- Loctagon Vial Skirt provides stability in freestanding position and locks into CryoELITE Benchmate Rack (985810) in order to provide easy open and close with one hand
- Exacting round bottom allows for up to 17,000 MAX RCF (xG)
- Supplied with caps attached
Legal Information
CryoELITE is a registered trademark of DWK LIFE SCIENCES LLC
Loctagon is a trademark of DWK LIFE SCIENCES LLC
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service