Skip to Content
Merck
All Photos(1)

Key Documents

D804

Sigma-Aldrich

γ-Decanolactone

98%

Synonym(s):

4-Hydroxydecanoic acid γ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.449 (lit.)

density

0.948 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC1CCC(=O)O1

InChI

1S/C10H18O2/c1-2-3-4-5-6-9-7-8-10(11)12-9/h9H,2-8H2,1H3

InChI key

IFYYFLINQYPWGJ-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hirotoshi Tamura et al.
Journal of agricultural and food chemistry, 53(13), 5397-5401 (2005-06-23)
Authenticity assessment of gamma-decalactone (1) and delta-decalactone (2) from peach (Prunus persica var. persica), apricot (Prunus armeniaca), and nectarine (Prunus persica var. nectarina) was performed using gas chromatography-isotope ratio mass spectrometry (GC-IRMS) in the combustion (C) and pyrolysis (P) mode.
Markus Lübke et al.
Journal of agricultural and food chemistry, 50(24), 7094-7099 (2002-11-14)
Interactions between a well-characterized protein, beta-lactoglobulin, and two flavor compounds, beta-ionone and gamma-decalactone, were studied by 2D NMR spectroscopy. NMR spectra were recorded in aqueous solution (pH 2.0, 12 mM NaCl, 10% D(2)O) under conditions such that beta-lactoglobulin is present
Mohamed Alchihab et al.
Journal of industrial microbiology & biotechnology, 37(2), 167-172 (2009-11-11)
During the biotransformation of castor oil into gamma-decalactone, R. aurantiaca produced both the lactone form and its precursor (4-hydroxydecanoic acid). After six days of culture, a maximum yield of gamma-decalactone of 6.5 g/l was obtained. The parameters of gamma-decalactone adsorption
Paulo Alexandre de Oliveira et al.
Experimental brain research, 187(1), 161-166 (2008-02-06)
Gamma-decanolactone is a monoterpene compound, and its psychopharmacological evaluation in mice revealed that it has a dose-dependent effect on the central nervous system, with hypnotic, anticonvulsant, and hypothermic activity. The aim of the present study was to investigate the effect
Mohamed Alchihab et al.
Applied biochemistry and biotechnology, 158(1), 41-50 (2008-07-22)
Among 18 psychrophilic strains isolated near the Antarctic Station, the psychrophilic strain Rhodotorula aurantiaca A19 was selected for its ability of growth and gamma-decalactone production at low temperatures. The effects of temperature, initial pH, and castor oil concentration on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service