Skip to Content
Merck
All Photos(1)

Key Documents

D7151

Sigma-Aldrich

1,7-Dioxaspiro[5.5]undecane

≥97%

Synonym(s):

Olive-fly ketal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

form

liquid

refractive index

n20/D 1.464 (lit.)

bp

193 °C/750 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CCC2(CCCCO2)OC1

InChI

1S/C9H16O2/c1-3-7-10-9(5-1)6-2-4-8-11-9/h1-8H2

InChI key

GBBVHDGKDQAEOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pheromone for Dacus oleae

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abby Jones Weldon et al.
The Journal of organic chemistry, 71(24), 9212-9216 (2006-11-18)
The relative gas-phase energetics of several low-lying isomers of 1,7-dioxaspiro[5.5]undecane and 1,7,9-trioxadispiro[5.1.5.3]hexadecane have been calculated with second-order Mller-Plesset perturbation theory and basis sets as large as aug-cc-pVQZ. Relative energies in THF, dichloromethane, acetone, and DMSO have been estimated with corrections
Anat Levi-Zada et al.
Journal of chemical ecology, 38(8), 1036-1041 (2012-09-04)
The olive fruit fly, Bactrocera oleae (Diptera: Tephritidae), uses 1,7-dioxaspiro[5.5]undecane ("olean"), produced primarily by females, as a sex pheromone. We used sequential solid phase microextraction-gas chromatography mass spectrometry (SPME-GCMS) analysis to show that female olive flies release about 1000 ng
S Makedonopoulou et al.
Acta crystallographica. Section B, Structural science, 57(Pt 3), 399-409 (2001-05-25)
The enantiomers of racemic olive fly sex pheromone 1,7-dioxaspiro[5.5]undecane (1) have been isolated by crystallization with enantiospecific cyclodextrin hosts: (S)-(1) crystallizes with heptakis(2,3,6-tri-O-methyl)-beta-cyclodextrin (TMbetaCD) and (R)-(1) with hexakis(2,3,6-tri-O-methyl)-alpha-cyclodextrin (TMalphaCD). The crystal structure of TMbetaCD/(S)-(1) from synchrotron radiation data at 100
Mary T Fletcher et al.
Chemical communications (Cambridge, England), (12)(12), 1302-1303 (2002-07-12)
The demonstration that both oxygen atoms of 1,7-dioxaspiro[5.5]undecane (1), the sex-pheromone of the female olive fly, originate from dioxygen, strongly implicates monooxygenase mediated processes in assembly of (1), and reveals unexpected complexity in the formation of its nine-carbon precursor.
N Rysanek et al.
Acta crystallographica. Section C, Crystal structure communications, 52 ( Pt 11), 2932-2936 (1996-11-15)
The complex beta-cyclodextrin-1,7-dioxaspiro[5,5]undecane nonahydrate, C42H70O35.C9H16O2.9H2O, belongs to the class of beta-cyclodextrin dimeric-type complexes. The racemic guest molecule is present in a disordered position. Both enantiomers are located in two different regions inside the channel formed by the host dimers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service