Skip to Content
Merck
All Photos(3)

Key Documents

140805

Sigma-Aldrich

1,4-Dibromobutane

99%

Synonym(s):

Tetramethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)4Br
CAS Number:
Molecular Weight:
215.91
Beilstein:
1071199
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

63-65 °C/6 mmHg (lit.)

mp

−20 °C (lit.)

density

1.808 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

BrCCCCBr

InChI

1S/C4H8Br2/c5-3-1-2-4-6/h1-4H2

InChI key

ULTHEAFYOOPTTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Dibromobutane reaction with atactic poly(2-vinylpyridine) leads to the formation of long-range 3D molecular ordering in polymer chains.

Application

1,4-Dibromobutane was used to investigate the metabolism of two halopropanes: 1,3-dichloropropane and 2,2-dichloropropane. It was used as reagent during the synthesis of diazadioxa oxovanadium(IV) macrocyclic complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Yu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 24(2), 162-165 (2005-03-17)
In phene-50% NaOH medium, 1,4-dibromobutane reacts with carbazole (CZ) to form N-bromobutylcarbaole (BCZ) at 70-80 degrees C. The 2-thenoyl-2-carbazolyl-N-butyltrifluoroacetone (TCBTA) was synthesized by reaction of BCZ with 2-thenoyltrifluoroacetone (TTA) in acetone-K2CO3 medium at 50 degrees C. The TCBTA is a
M L Sharma et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 95, 562-568 (2012-05-15)
A new series of diazadioxa oxovanadium(IV) macrocyclic complexes of type [VO(mac)]SO(4) have been synthesized via the condensation reaction of a 3-(phenyl/substituted phenyl)-4-amino-5-hydrazino-1,2,4-triazole (H(2)L) with salicylaldehyde/2-hydroxyacetophenone and 1,4-dibromobutane in the presence of oxovanadium(IV) sulfate in ethanol. All the newly synthesized compounds
Weijiang Guan et al.
Nature communications, 7, 11811-11811 (2016-06-03)
Inorganic dispersion is of great importance for actual implementation of advanced properties of organic-inorganic composites. Currently, electron microscopy is the most conventional approach for observing dispersion of inorganic fillers from ultrathin sections of organic-inorganic composites at the nanoscale by professional
Mange R Yadav et al.
ACS omega, 3(9), 11802-11816 (2018-10-16)
Some quaternary gemini amphiphiles (GAs) were synthesized as nonviral gene delivery carriers. The critical miceller concentration values of these amphiphiles are indicative of their superior surface-active properties. All of the synthesized GAs, alone or along with lipids like cholesterol and/or
W Onkenhout et al.
Drug metabolism and disposition: the biological fate of chemicals, 14(5), 608-612 (1986-09-01)
Two stable sulfur-containing metabolites were isolated from rat urine following administration of the mutagenic 1,4-dibromobutane. They were identified as tetrahydrothiophene and 3-hydroxysulfolane by gas chromatography and gas chromatography-mass spectrometry and were found to be excreted in 48-hr urine, representing 5.8

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service