All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H10Br2
CAS Number:
Molecular Weight:
241.95
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.5515 (lit.)
bp
145 °C/100 mmHg (lit.)
density
1.784 g/mL at 25 °C (lit.)
SMILES string
Br[C@@H]1CCCC[C@H]1Br
InChI
1S/C6H10Br2/c7-5-3-1-2-4-6(5)8/h5-6H,1-4H2/t5-,6-/m1/s1
InChI key
CZNHKZKWKJNOTE-PHDIDXHHSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service