Skip to Content
Merck
All Photos(1)

Key Documents

333549

Sigma-Aldrich

3-Bromocyclohexene

technical grade, 90%

Synonym(s):

2-Cyclohexen-1-yl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9Br
CAS Number:
Molecular Weight:
161.04
Beilstein:
635953
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.528 (lit.)

bp

57-58 °C/12 mmHg (lit.)

density

1.4 g/mL at 25 °C (lit.)

functional group

bromo

storage temp.

−20°C

SMILES string

BrC1CCCC=C1

InChI

1S/C6H9Br/c7-6-4-2-1-3-5-6/h2,4,6H,1,3,5H2

InChI key

AJKDUJRRWLQXHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Bromocyclohexene was used in the synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine. It was also used in the synthesis of enantiopure cyclohexitols such as muco-quercitol, D-chiro-inocitol and allo-inocitol.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Nishiyama et al.
Chemical & pharmaceutical bulletin, 49(2), 233-235 (2001-02-24)
A facile and efficient synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine [Boc-Tyr(Chx)-OH] is described. Boc-Tyr-OH was treated with NaH in dimethylformamide and then with 3-bromocyclohexene to give N-Boc-O-(cyclohex-2-enyl)-L-tyrosine [Boc-Tyr(Che)-OH] in 70% yield. Hydrogenation of Boc-Tyr(Che)-OH over PtO2 afforded Boc-Tyr(Chx)-OH in almost quantitative yield. The
Synthesis of enantiopure cyclitols from (?)-3-bromocyclohexene mediated by intramolecular oxyselenenylation employing (S, S)-hydrobenzoin and (S)-mandelic acid as chiral sources.
Lee YJ, et al.
Tetrahedron, 61(8), 1987-2001 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service