Skip to Content
Merck
All Photos(4)

Key Documents

471232

Sigma-Aldrich

Dibutylamine

≥99.5%

Synonym(s):

N,N-Di-n-butylamine, N-Butyl-1-butanamine, n-Dibutylamine, Di-n-butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2NH
CAS Number:
Molecular Weight:
129.24
Beilstein:
506001
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
bp:
159 °C (lit.)
vapor pressure:
1.9 mmHg ( 20 °C)

vapor density

4.46 (vs air)

vapor pressure

1.9 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

594 °F

expl. lim.

10 %

refractive index

n20/D 1.417 (lit.)

pH

11.1 (20 °C, 1 g/L)

bp

159 °C (lit.)

mp

−62 °C (lit.)

solubility

water: soluble 3.8 g/L at 20 °C

density

0.767 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCNCCCC

InChI

1S/C8H19N/c1-3-5-7-9-8-6-4-2/h9H,3-8H2,1-2H3

InChI key

JQVDAXLFBXTEQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutylamine is a highly reactive catalyst, commonly used as a reagent in the derivatization of isocyanates.

Application

Dibutylamine can be used as a reactant to synthesize:,·      
  • N, N




  • -Dibutyl-4-methylaniline by Pd-catalyzed Buchwald-Hartwig amination reaction with aryl chlorides and bromides.     
  • N,N-Dibutylacetamide by acetylation reaction with isopropenyl acetate.

It can also be used as:      
  • A strong hindered base in the different elimination reactions.
  • An organocatalyst to synthesize annulated pyrano[2,3-d]pyrimidine derivatives via multicomponent condensation reaction of aromatic aldehydes, malononitrile, and barbituric acid.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.9 °F - closed cup

Flash Point(C)

40.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of complex mixtures of airborne isocyanates and amines. Part 1. Liquid chromatography with ultraviolet detection of monomeric and polymeric isocyanates as their dibutylamine derivatives.
Spanne M, et al.
Analyst, 121(8), 1095-1099 (1996)
Chemistry of blocked isocyanates. II. Kinetics and mechanism of the reaction of dibutylamine with phenyl and 2-methylphenyl oxime carbamates.
Levine AW and Fech Jr J.
The Journal of Organic Chemistry, 37(15), 2455-2460 (1972)
Dibutylamine (DBA): A highly efficient catalyst for the synthesis of pyrano [2, 3-d] pyrimidine derivatives in aqueous media
Ajmal R B et al.
Journal of Taibah University for Science , 10, 9-18 (2016)
Determination of complex mixtures of airborne isocyanates and amines. Part 3. Methylenediphenyl diisocyanate, methylenediphenylamino isocyanate and methylenediphenyldiamine and structural analogues after thermal degradation of polyurethane
Haakan T et al.
Analyst, 122, 275-278 (1997)
Julien Peyrton et al.
Molecules (Basel, Switzerland), 24(23) (2019-12-01)
Nowadays, polyols are basic chemicals for the synthesis of a large range of polymers, such as polyurethane foams (PUF), which are produced with several other compounds, such as polyisocyanates. During the last decades, the oleo-chemistry has developed several routes from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service