Skip to Content
Merck
All Photos(1)

Key Documents

D131806

Sigma-Aldrich

3,4-Dimethoxybenzoic acid

≥99%

Synonym(s):

Veratric acid, 3,4-Dimethoxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CO2H
CAS Number:
Molecular Weight:
182.17
Beilstein:
518285
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

powder

mp

179-182 °C (lit.)

SMILES string

COc1ccc(cc1OC)C(O)=O

InChI

1S/C9H10O4/c1-12-7-4-3-6(9(10)11)5-8(7)13-2/h3-5H,1-2H3,(H,10,11)

InChI key

DAUAQNGYDSHRET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shicheng Chen et al.
FEMS microbiology letters, 218(1), 143-148 (2003-02-14)
Volvariella volvacea, strain V14, produces multiple forms of extracellular laccase when grown in submerged culture in a defined medium with glucose as sole carbon source, and on cotton waste 'compost' representative of the conditions used for industrial-scale mushroom cultivation. In
Motohiro Nishimura et al.
Bioscience, biotechnology, and biochemistry, 70(9), 2316-2319 (2006-09-09)
The vanillate demethylase genes from Streptomyces sp. NL15-2K were cloned and sequenced. The vanA and vanB gene homologs, which encode the terminal oxygenase subunit (VanA) and the ferredoxin-type reductase subunit (VanB) of the enzyme respectively, were found in the sequenced
Gary Ward et al.
The Journal of biological chemistry, 278(41), 39726-39734 (2003-07-15)
The steady state kinetic parameters Km and kcat for the oxidation of phenolic substrates by lignin peroxidase correlated with the presteady state kinetic parameters Kd and k for the reaction of the enzyme intermediate compound II with the substrates, indicating
Jian-Hua Zou et al.
Journal of natural products, 67(4), 664-667 (2004-04-24)
Six new acylated flavone C-glycosides, 2''-O-(2'''-methylbutyryl)isoswertisin (1), 3''-O-(2'"-methylbutyryl)isoswertisin (2), 2''-O-(2'"-methylbutyryl)vitexin (3), 2''-O-(2'"-methylbutyryl)orientin (4), 2''-O-(3'",4'"-dimethoxybenzoyl)vitexin (5), and 2''-O-(3'",4'"-dimethoxybenzoyl)orientin (6), and the known flavone C-glycoside, isoswertisin, were isolated from the flowers of Trollius ledebouri. The structures of 1-6 were elucidated by spectroscopic
Pedro A R Fernandes et al.
Carbohydrate polymers, 223, 115132-115132 (2019-08-21)
In this work, polysaccharides extracted with hot water from apple pomace were isolated by C18 cartridge solid-phase extraction at pH 7 (Fr7). Dialysis (12-14 kDa) of this fraction allowed to obtain 17% (w/w) of polymeric material composed by 65% of polysaccharides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service