Skip to Content
Merck
All Photos(1)

Key Documents

156639

Sigma-Aldrich

3′,4′-Dimethoxyacetophenone

98%

Synonym(s):

1-(3,4-Dimethoxyphenyl)ethan-1-one, 1-(3,4-Dimethoxyphenyl)ethanone, 1-(4,5-Dimethoxyphenyl)ethanone, 2-Fluoro-1-nitrobenzene, 2-Nitro-1-fluorobenzene, 3,4-Dimethoxyphenyl methyl ketone, 3′,4′-Dimethoxyacetophenone, 4-Acetyl-1,2-dimethoxybenzene, 4-Acetylveratrole, Methyl 3,4-dimethoxyphenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3COCH3
CAS Number:
Molecular Weight:
180.20
Beilstein:
781213
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

286-288 °C (lit.)

mp

47-54 °C (lit.)

solubility

hot water: soluble

functional group

ketone

SMILES string

COc1ccc(cc1OC)C(C)=O

InChI

1S/C10H12O3/c1-7(11)8-4-5-9(12-2)10(6-8)13-3/h4-6H,1-3H3

InChI key

IQZLUWLMQNGTIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service