Skip to Content
Merck
All Photos(1)

Key Documents

CDS000103

Sigma-Aldrich

2-Hexadecenoic acid

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30O2
Molecular Weight:
254.41
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

form

solid

SMILES string

CCCCCCCCCCCCC\C=C\C(O)=O

InChI

1S/C16H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h14-15H,2-13H2,1H3,(H,17,18)/b15-14+

InChI key

ZVRMGCSSSYZGSM-CCEZHUSRSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mohammad Norazmi Ahmad et al.
Scientific reports, 10(1), 9566-9566 (2020-06-14)
Lipid oxidation and microbial contamination are the major factors contributing to food deterioration. Food additives like antioxidants and antibacterials can prevent food spoilage by delaying oxidation and preventing the growth of bacteria. Artocarpus altilis leaves exhibited biological properties that suggested
Hong-Ying Gao et al.
Journal of inflammation (London, England), 9(1), 16-16 (2012-05-09)
The aim of this study was to investigate the hepatoprotective effect of Matrine salvianolic acid B salt on carbon tetrachloride (CCl4)-induced hepatic fibrosis in rats. Salvianolic acid B and Matrine has long been used to treat liver fibrosis. Matrine salvianolic
Dan Ouyang et al.
The Analyst, 145(8), 3125-3130 (2020-03-13)
A spherical vinyl-functionalized covalent-organic framework (COF-V) was prepared at room temperature by a facile method and applied as a novel substrate for surface-enhanced laser desorption/ionization mass spectrometry (SELDI-MS). Compared with conventional organic matrices, the spherical COF-V with high crystallinity and
Christiane Theiss et al.
Journal of pharmaceutical and biomedical analysis, 160, 212-221 (2018-08-14)
Macrogol-based emulsifiers and their respective precursor substances, i.e. macrogols (PEG), fatty acids (FA), and fatty alcohols (FAA), are widely used excipients which are usually characterized by a series of tests described within the European Pharmacopoeia (Ph. Eur.). Examples are bulk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service