Merck
  • A spherical covalent-organic framework for enhancing laser desorption/ionization mass spectrometry for small molecule detection.
A spherical covalent-organic framework for enhancing laser desorption/ionization mass spectrometry for small molecule detection.
The Analyst (2020-03-13)
Dan Ouyang, Kailong Luo, Wende Ma, Jie Wu, Jing Li, Yanting He, Zongwei Cai, Zian Lin
ABSTRACT
A spherical vinyl-functionalized covalent-organic framework (COF-V) was prepared at room temperature by a facile method and applied as a novel substrate for surface-enhanced laser desorption/ionization mass spectrometry (SELDI-MS). Compared with conventional organic matrices, the spherical COF-V with high crystallinity and good monodispersity exhibited high sensitivity, no matrix background interference, wide-range applicability, high salt tolerance and reproducibility in the characterization of small molecules. Considering these advantages, the applicability of the spherical COF-V-based SELDI-MS method was successfully demonstrated by determining trace amounts of glucose in diabetic urine, which would be a promising candidate for clinical diagnosis of diabetes. In addition, the morphological effect and the desorption/ionization mechanism of the COF-V were investigated in detail and the results indicated that the spherical COF-V substrate could greatly enhance the LDI process compared with the bulk COF-V. This work not only extends the application of COFs in MS, but also offers a promising alternative for small molecule identification and clinical diagnosis of diabetes.
Sigma-Aldrich
Bis(4-hydroxyphenyl)methane, 98%
Supelco
9-Aminoacridine, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
Maltose solution, for molecular biology, BioReagent, ~20% in H2O