Skip to Content
Merck
All Photos(1)

Key Documents

CDS000009

Sigma-Aldrich

6-Amino-9H-purin-2-ol

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5O
CAS Number:
Molecular Weight:
151.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

Nc1nc(O)nc2[nH]cnc12

InChI

1S/C5H5N5O/c6-3-2-4(8-1-7-2)10-5(11)9-3/h1H,(H4,6,7,8,9,10,11)

InChI key

DRAVOWXCEBXPTN-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Kawai et al.
Life sciences in space research, 20, 20-29 (2019-02-25)
Titan is the largest moon of Saturn and possesses a dense atmosphere composed of nitrogen and methane. Various types of organic compounds (hydrocarbons, nitriles, etc.) have been found on Titan, which were generated by reactions taking place in its atmosphere.
Shu R Huang et al.
The journal of physical chemistry. A, 124(35), 7101-7112 (2020-08-14)
Oxidation of nontraditional nucleobases 1-methylcytosine (hachimoji base S) and isoguanine (hachimoji base B) in gas-phase ternary complexes with CuII(terpyridine)2+ formed cation radicals that were characterized by tandem mass spectrometry, UV-vis photodissociation action spectroscopy in the 210-700 nm region, and ab
Adam M Weidner et al.
PloS one, 6(9), e24930-e24930 (2011-09-29)
While research supports amyloid-β (Aβ) as the etiologic agent of Alzheimer's disease (AD), the mechanism of action remains unclear. Evidence indicates that adducts of RNA caused by oxidation also represent an early phenomenon in AD. It is currently unknown what
Christopher K Materese et al.
Astrobiology, 17(8), 761-770 (2017-07-21)
Nucleobases are the informational subunits of RNA and DNA and are essential to all known forms of life. The nucleobases can be divided into two groups of molecules: the pyrimidine-based compounds that include uracil, cytosine, and thymine, and the purine-based
Sam Dawbaa et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1051, 84-91 (2017-03-24)
Oxidative stress is considered as one of the significant causes of DNA damage which in turn contributes to cell death through a series of intermediate processes such as cancer formation, mutation, and aging. Natural sources such as plant and fruit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service