Skip to Content
Merck
All Photos(1)

Key Documents

C99701

Sigma-Aldrich

Bromocycloheptane

97%

Synonym(s):

Cycloheptyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13Br
CAS Number:
Molecular Weight:
177.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.5052 (lit.)

bp

71-72 °C/10 mmHg (lit.)

density

1.289 g/mL at 25 °C (lit.)

SMILES string

BrC1CCCCCC1

InChI

1S/C7H13Br/c8-7-5-3-1-2-4-6-7/h7H,1-6H2

InChI key

LOXORFRCPXUORP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service