B77207
1-Bromo-3-phenylpropane
98%
Synonym(s):
(3-Bromopropyl)benzene, 3-Phenylpropyl bromide, Hydrocinnamyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5(CH2)3Br
CAS Number:
Molecular Weight:
199.09
Beilstein:
2205527
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.546 (lit.)
bp
237-238 °C (lit.)
density
1.31 g/mL at 25 °C (lit.)
SMILES string
BrCCCc1ccccc1
InChI
1S/C9H11Br/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2
InChI key
XMZQWZJMTBCUFT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
1-Bromo-3-phenylpropane is used in the etherification reaction under phase-transfer catalytic conditions.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
215.6 °F - closed cup
Flash Point(C)
102 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Swen Schuster et al.
Physical chemistry chemical physics : PCCP, 21(18), 9098-9105 (2019-04-25)
Photoisomerization of a series of custom-designed, azobenzene-substituted alkanethiolate (AT) self-assembled monolayers (SAMs) on Au(111) substrates was studied in the context of work function variation, using Kelvin probe measurements as a transduction technique. These SAMs featured variable packing density (by ∼14%;
Kinetic study of synthesizing 1-phenyl-3-propyl ether under liquid-liquid phase-transfer catalytic conditions
Maw-Ling W et al.
Chemical Engineering Communications , 196, 573-590 (2008)
Xiaojie Zhao et al.
Journal of chromatography. A, 1587, 197-208 (2018-12-26)
Ionic liquid bonded polysiloxanes (PILs) are a class of polysiloxanes whose side chains contain ionic liquid (IL) moieties. Considering their excellent selectivity and thermo-stability, PILs have great potentials in the development of polar stationary phases for gas chromatography. In this
Jiang Yan et al.
Science advances, 6(45) (2020-11-08)
Phenol is an important commodity chemical in the industry, which is currently produced using fossil feedstocks. Here, we report a strategy to produce phenol from lignin by directly deconstructing Csp2-Csp3 and C-O bonds under mild conditions. It was found that
Hayrettin O Gülcan et al.
Archiv der Pharmazie, 348(1), 55-61 (2015-01-13)
The current structure-activity relationship of profens (i.e., 2-arylpropionic acid derivatives, a class of non-steroidal anti-inflammatory drugs) discusses the importance of α-monomethyl substitution on these compounds, since the activities obtained through their corresponding arylacetic acid derivatives (i.e., α-demethylated derivatives) or α,α-dimethyl-substituted
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service