C95706
Cyclobutanecarbonyl chloride
98%
Synonym(s):
Cyclobutancarboxylic acid chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C4H7COCl
CAS Number:
Molecular Weight:
118.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.455 (lit.)
bp
60 °C/50 mmHg (lit.)
density
1.039 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
ClC(=O)C1CCC1
InChI
1S/C5H7ClO/c6-5(7)4-2-1-3-4/h4H,1-3H2
InChI key
JFWMYCVMQSLLOO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
98.6 °F - closed cup
Flash Point(C)
37 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service