Skip to Content
Merck
All Photos(1)

Key Documents

C92804

Sigma-Aldrich

1-Cyanonaphthalene

98%

Synonym(s):

1-Naphthonitrile, 1-Cyanonaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CN
CAS Number:
Molecular Weight:
153.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

bp

299 °C (lit.)

mp

36-38 °C (lit.)

SMILES string

N#Cc1cccc2ccccc12

InChI

1S/C11H7N/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H

InChI key

YJMNOKOLADGBKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service