Skip to Content
Merck
All Photos(1)

Key Documents

B56609

Sigma-Aldrich

9-Bromoanthracene

94%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Br
CAS Number:
Molecular Weight:
257.13
Beilstein:
1869747
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

94%

form

crystals

mp

97-100 °C (lit.)

SMILES string

Brc1c2ccccc2cc3ccccc13

InChI

1S/C14H9Br/c15-14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9H

InChI key

ZIRVQSRSPDUEOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengbo Xie et al.
Luminescence : the journal of biological and chemical luminescence, 33(3), 604-610 (2018-01-30)
With carbazole and p-cyanobromobenzene as raw materials, 4-(3,6-di (anthracen-9-yl)-9H-carbazol-9-yl)benzonitrile (DACB) and 4-(3,6-bis(anthracene -9-ylethynyl)-9H-carbazol-9-yl)benzonitrile (BACB) were synthesized through the Suzuki coupling reaction and the Sonogashira coupling reaction, respectively. These structures were characterized using 1 H nuclear magnetic resonance (NMR), elemental analysis
Irena Bylińska et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 15(1), 45-56 (2015-12-01)
A series of symmetrically substituted diarylacetylenes and diaryl-1,3-butadiynes were prepared and studied with an emphasis on their spectral and photophysical properties. The photophysical characteristics of these compounds were studied in relation to their structures and the influence of solvent or
Rong Jin et al.
Journal of chromatography. A, 1509, 114-122 (2017-06-18)
Isotopic dilution gas chromatography combined with high resolution mass spectrometry (GC/HRMS) has overwhelming advantages with respect to the accuracy of congener-specific ultratrace analysis of complex persistent organic pollutants (POPs) in environmental matrices. However, an isotopic dilution GC/HRMS method for analysis
Jian-Lin Sun et al.
Ecotoxicology and environmental safety, 113, 31-37 (2014-12-09)
Halogenated polycyclic aromatic hydrocarbons (HPAHs) have been reported to occur widely in urban air. Nevertheless, knowledge about the human health risk associated with inhalation exposure to HPAHs is scarce so far. In the present study, nine HPAHs and 16 PAHs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service