Merck
  • Sodium-activated potassium channels are functionally coupled to persistent sodium currents.
Sodium-activated potassium channels are functionally coupled to persistent sodium currents.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2012-02-24)
Travis A Hage, Lawrence Salkoff
ABSTRACT
We report a novel coupled system of sodium-activated potassium currents (I(KNa)) and persistent sodium currents (I(NaP)), the components of which are widely distributed throughout the brain. Its existence and importance has not been previously recognized. Although I(KNa) was known to exist in many cell types, the source of Na(+) which activates I(KNa) remained a mystery. We now show in single membrane patches generated from the somas of rat neurons that sodium influx through I(NaP) is sufficient for activation of K(Na) channels, without substantial contribution from the transient sodium current or bulk [Na(+)](i). I(NaP) was found to be active at cell membrane resting potentials, a finding that may explain why I(KNa) can be evoked from negative holding potentials. These results show an unanticipated role for I(NaP) in activating a negative feedback system countering the excitable effects I(NaP); the interrelatedness of I(NaP) and I(KNa) suggests new ways neurons can tune their excitability.
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥98%
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
Sigma-Aldrich
Veratridine, ≥90% (HPLC), powder
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis
Sigma-Aldrich
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis
Sigma-Aldrich
Cesium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cesium chloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Cesium chloride, Grade II, ≥98%
Sigma-Aldrich
Cesium chloride, ReagentPlus®, 99.9%
Sigma-Aldrich
Cesium chloride, puriss. p.a., ≥99.5%