Skip to Content
Merck
All Photos(2)

Key Documents

235806

Sigma-Aldrich

3,4-Dichlorobenzotrifluoride

97%

Synonym(s):

3,4-Dichloro-α,α,α-trifluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CF3
CAS Number:
Molecular Weight:
215.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.6 mmHg ( 20 °C)

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.475 (lit.)

bp

173-174 °C (lit.)

mp

−13-−12 °C (lit.)

density

1.478 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccc(Cl)c(Cl)c1

InChI

1S/C7H3Cl2F3/c8-5-2-1-4(3-6(5)9)7(10,11)12/h1-3H

InChI key

XILPLWOGHPSJBK-UHFFFAOYSA-N

Application

3,4-Dichlorobenzotrifluoride has been used in the preparation of:
  • 3,4-difluorobenzotrifluoride by a KF exchange reaction[1]
  • 2-chloro-4-trifluoromethylphenyl 4-nitrophenylether (RH-2512), potential diphenyl ether herbicide[2]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New potent diphenyl ether herbicides.
R Y Yih et al.
Journal of agricultural and food chemistry, 23(3), 592-593 (1975-05-01)
Aromatic fluorine chemistry. Part 1. 3, 4-Difluorobenzoic acid and derivatives via 3, 4-difluorobenzotrifluoride.
Pews RG, et al.
Journal of Fluorine Chemistry, 50(3), 365-370 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service