Skip to Content
Merck
All Photos(3)

Key Documents

C106402

Sigma-Aldrich

N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide methyl-p-toluenesulfonate

95%

Synonym(s):

4-[2-[(Cyclohexylcarbonimidoyl)amino]ethyl]-4-methylmorpholinium p-toluenesulfonate, N-Cyclohexyl-N′-(β-[N-methylmorpholino]ethyl)carbodiimide p-toluenesulfonate salt, CMC, CMC metho-p-toluenesulfonate, Morpho CDI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26N3O · C7H7O3S
CAS Number:
Molecular Weight:
423.57
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

95%

form

crystals

mp

113-115 °C (lit.)

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.C[N+]2(CCOCC2)CCN=C=NC3CCCCC3

InChI

1S/C14H26N3O.C7H8O3S/c1-17(9-11-18-12-10-17)8-7-15-13-16-14-5-3-2-4-6-14;1-6-2-4-7(5-3-6)11(8,9)10/h14H,2-12H2,1H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1

InChI key

GBCAVSYHPPARHX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Employed in a facile sequencing technique for pseudouridylate residues in RNA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Bakin et al.
Biochemistry, 32(37), 9754-9762 (1993-09-21)
A new technique has been developed for the facile location of pseudouridylate (psi) residues in any RNA molecule. The method uses two known modification procedures which in combination uniquely identify U residues which have been converted into psi. The first
Carola Schafer et al.
Malaria journal, 17(1), 370-370 (2018-10-20)
Plasmodium vivax is the most geographically widespread of the human malaria parasites, causing 50,000 to 100,000 deaths annually. Plasmodium vivax parasites have the unique feature of forming dormant liver stages (hypnozoites) that can reactivate weeks or months after a parasite-infected
S L Larsson et al.
Biochemistry, 40(10), 3222-3231 (2001-03-22)
The expansion segments in eukaryotic ribosomal RNAs are additional RNA sequences not found in the RNA core common to both prokaryotes and eukaryotes. These regions show large species-dependent variations in sequence and size. This makes it difficult to create secondary
K G Patteson et al.
Nucleic acids research, 29(10), E49-E49 (2001-05-23)
A new method using matrix-assisted laser desorption/ionization (MALDI) mass spectrometry for the direct analysis of the mass-silent post-transcriptionally modified nucleoside pseudouridine in nucleic acids has been developed. This method utilizes 1-cyclohexyl-3-(2-morpholinoethyl)carbodiimide to derivatize pseudouridine residues. After chemical derivatization all pseudouridine
Jennifer M Bailey et al.
Journal of virology, 81(2), 650-668 (2006-11-03)
Coxsackievirus B3 (CVB3) is a picornavirus which causes myocarditis and pancreatitis and may play a role in type I diabetes. The viral genome is a single 7,400-nucleotide polyadenylated RNA encoding 11 proteins in a single open reading frame. The 5'

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service