Key Documents
BR91273
BRAND® glass beaker with spout, low form
volume 5000 mL, with graduation
Synonym(s):
Beakers, Glass
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
borosilicate glass 3.3
feature
with graduation
manufacturer/tradename
BRAND 91273
volume
5000 mL
General description
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards, this low-form beaker features clear graduation markings for precise measurement of liquids. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer.
Other Notes
Browse all glass beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)