BR782730
BRAND® 12-well Cell Culture Insert, PET-membrane
pore size 0.4 μm, pack of 48 pieces
All Photos(1)
About This Item
Recommended Products
material
PET membrane
packaging
pack of 48 pieces
manufacturer/tradename
BRAND 782730
size
(12 well)
pore size
0.4 μm
General description
The BRAND Insert 2in1 is a cell culture insert used for skin model production, 3D epithelium cultures, chemotaxis analyses, transmigration assays, transport and polarity studies, and more.
In many cell culture tasks, or when switching from a standing to a hanging position, you can continue to use your current cell culture plates, saving additional validation steps. Thanks to both supporting feet and hooks, you can position the BRAND Insert 2in1 either way.
Via its protein-like structure, the cellGrade™ plus surface provides optimal conditions for growing primary keratinocytes and many other cell types.
With the practical multipacks, the right number of inserts is always close at hand. They can be opened row by row, so that the remaining inserts stay sterile. Besides the practical benefits, this also reduces waste.
Overview
In many cell culture tasks, or when switching from a standing to a hanging position, you can continue to use your current cell culture plates, saving additional validation steps. Thanks to both supporting feet and hooks, you can position the BRAND Insert 2in1 either way.
Via its protein-like structure, the cellGrade™ plus surface provides optimal conditions for growing primary keratinocytes and many other cell types.
With the practical multipacks, the right number of inserts is always close at hand. They can be opened row by row, so that the remaining inserts stay sterile. Besides the practical benefits, this also reduces waste.
Overview
- Use in a hanging or standing position
- Works with all common 6-, 12-, or 24-well multi-well plates
- Suitable for 2D and 3D cell cultures
- Surface: cellGrade™ plus
- Sterility Assurance Level (SAL) of 10-6
- PC or PET membrane
- Pore size: from 0.4 μm and 8.0 μm
- Packaged individually or in practical multi-packs
Features and Benefits
• Inserts can be used in a hanging or standing position• For 24-, 12-, and 6-well plates or culture dishes• Uniform and controlled cell growth thanks to surface treatment
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
Already Own This Product?
Articles
The BRAND® Insert 2in1 cell culture insert supports the cultivation of Reconstructed Human Epidermis used for skin corrosion tests.