BR466134
BRAND® filter funnel
borosilicate glass, capacity 500 mL, 25 D 4
Synonym(s):
Filter, Filter funnel
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
manufacturer/tradename
BRAND 466134
capacity
500 mL
filter diam.
9.5 cm
pore size
10-16 μm
Looking for similar products? Visit Product Comparison Guide
General description
Crafted from borosilicate glass, known for its excellent thermal and chemical resistance, making it suitable for a wide range of substances.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service