BR36172
BRAND® volumetric flask
PMP, transparent, size 25 mL
Synonym(s):
Plastics, Volumetric flasks
All Photos(1)
About This Item
Recommended Products
material
PMP
polypropylene stopper
transparent
Joint
joint ST/NS10/19
feature
accuracy class A
packaging
pack of 1 ea
manufacturer/tradename
BRAND 36172
size
25 mL
General description
Error limits according to class A, DIN EN ISO 1042.
Calibrated to contain (TC, In). Exposure to temperatures up to 121 °C (autoclaving) will not cause permanently exceeded error limits!
Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions.
Calibrated to contain (TC, In). Exposure to temperatures up to 121 °C (autoclaving) will not cause permanently exceeded error limits!
Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)