BR146519
BRAND® glass powder funnel short wide stem
top diam. 100 mm, bottom diam. 25 mm
Synonym(s):
Funnels, Power funnels
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
BRAND 146519
bottom diam.
25 mm
top diam.
100 mm
General description
The BRAND® Glass Powder Funnel with Short Wide Stem is expertly crafted from Boro 3.3 glass, ensuring durability and high performance in laboratory applications. This funnel features a ground and fire-polished upper rim, providing a smooth edge for safe handling and easy pouring. Designed with a short, wide stem, it facilitates efficient transfer of powders and granular materials, minimizing the risk of clogs and ensuring a swift flow.
Features and Benefits
- BRAND® borosilicate glass powder funnel is best suited for transferring powders.
- These general lab use funnels include ground and fire-polished upper rim.
- Features a short, wide stem that facilitates optimal filtration.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)