Skip to Content
Merck
All Photos(1)

Key Documents

B80208

Sigma-Aldrich

3-Bromopyridine

99%

Synonym(s):

3-Bromopyridine, 3-Pyridyl bromide, 5-Bromopyridine, Pyridin-3-yl bromide, m-Bromopyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4BrN
CAS Number:
Molecular Weight:
158.00
Beilstein:
105880
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.571 (lit.)

bp

173 °C (lit.)

density

1.64 g/mL at 25 °C (lit.)

SMILES string

Brc1cccnc1

InChI

1S/C5H4BrN/c6-5-2-1-3-7-4-5/h1-4H

InChI key

NYPYPOZNGOXYSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dylan J Walsh et al.
Nature communications, 11(1), 3094-3094 (2020-06-20)
The properties of a polymer are known to be intrinsically related to its molecular weight distribution (MWD); however, previous methodologies of MWD control do not use a design and result in arbitrary shaped MWDs. Here we report a precise design
Jeyeong Lee et al.
Dalton transactions (Cambridge, England : 2003), 49(42), 15002-15008 (2020-10-24)
The self-assembly of Pd(NO3)2 with L (L = 1,2-bis(dimethyl(pyridin-3-yl)silyl)ethane) gives rise to [PdL2](NO3)2 in high yields. Anion exchange of [PdL2](NO3)2 with X- (X- = BF4-, ClO4-, and PF6-) changes the skeleton into a cage of [(X)@Pd2L4](X)3. Successive anion exchange of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service