Skip to Content
Merck
All Photos(1)

Key Documents

558761

Sigma-Aldrich

2-Iodopyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4IN
CAS Number:
Molecular Weight:
205.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.6320 (lit.)

bp

52 °C (lit.)

density

1.928 g/mL at 25 °C (lit.)

functional group

iodo

SMILES string

Ic1ccccn1

InChI

1S/C5H4IN/c6-5-3-1-2-4-7-5/h1-4H

InChI key

CCZWSTFVHJPCEM-UHFFFAOYSA-N

General description

2-Iodopyridine can be synthesized from 2-chloropyridine or 2-bromopyridine via treatment with iodotrimethylsilane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational assignments of 2-iodopyridine.
Sortur V, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 69(2), 604-611 (2008)
Lianzhong Luo et al.
Environmental toxicology and chemistry, 36(10), 2602-2613 (2017-03-18)
Oysters accumulate Zn as an adaptation to Zn exposure; however, it is not known whether male and female oysters respond differently to Zn exposure. Proteomic and real-time polymerase chain reaction analyses were used to investigate differential responses of male and
"The solid complex Zn (CF 3) Br? 2DMF as an alternative reagent for the preparation of both, trifluoromethyl and pentafluoroethyl copper, CuCF3 and CuCF5"
Kremlev MM, et al.
Journal of Fluorine Chemistry, 131(2), 212-216 (2010)
Tri-(2-pyridyl) amine.
Wibaut JP and La Bastide GLC.
Rec. Trav. Chim., 52(6), 493-498 (1993)
Photo-arylation. IV. Synthesis of 2-arylpyridines by photo-reaction of 2-iodopyridine with substituted benzenes.
Terashima M, et al.
Chemical & Pharmaceutical Bulletin, 33(3), 1009-1015 (1985)

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service