Skip to Content
Merck
All Photos(1)

Key Documents

124036

Sigma-Aldrich

1-Bromo-3-chlorobenzene

99%

Synonym(s):

1-Chloro-3-bromobenzene, 3-Bromo-1-chlorobenzene, 3-Chloro-1-bromobenzene, 3-Chlorophenyl bromide, m-Bromochlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4Cl
CAS Number:
Molecular Weight:
191.45
Beilstein:
1446353
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39080308
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.576 (lit.)

bp

196 °C (lit.)

density

1.63 g/mL at 25 °C (lit.)

functional group

bromo
chloro

SMILES string

Clc1cccc(Br)c1

InChI

1S/C6H4BrCl/c7-5-2-1-3-6(8)4-5/h1-4H

InChI key

JRGGUPZKKTVKOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-3-chlorobenzene undergoes bromine-magnesium exchange reactions with i-PrMgCl-LiCl in THF at 0°C and is strongly accelerated by electron-withdrawing substituents. It acts as test substrate to study the application of a new catalytic system for cyanation of various aryl halides using K4[Fe(CN)6] as cyanating source.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Shi et al.
The Journal of organic chemistry, 74(7), 2760-2764 (2009-03-14)
Competition experiments have been performed to determine the relative reactivities of substituted bromobenzenes, bromonaphthalenes, and 9-bromoanthracene toward i-PrMgCl x LiCl in THF at 0 degrees C. The rates of the bromine-magnesium exchange reactions are accelerated by electron-acceptor substituents, the activating
K Schümann et al.
European journal of clinical investigation, 34(4), 275-282 (2004-04-17)
Iron regulatory protein 1 (IRP1), a post-transcriptional regulator of iron metabolism, is activated in the duodenum of iron-deficient animals, which is associated with increased iron absorption. In cell cultures IRP1 was also activated by iron-independent signals, such as H(2)O(2). Here
An efficient palladium catalytic system for microwave assisted cyanation of aryl halides.
Hajipour AR, et al.
Journal of Organometallic Chemistry, 696(4), 819-824 (2011)
Silvina C Pérez et al.
Solid state nuclear magnetic resonance, 87, 18-23 (2017-07-01)
A simple model to explain the NQR lineshape in solids with orientational disorder or substitutional disorder is presented. The particular case of m-chlorobromobenzene is studied. It is based on the assumption that Bromine atoms, of m-chlorobromobenzene molecules, behave as point
Nicholas C Pflug et al.
Environmental science & technology, 53(9), 4813-4822 (2019-03-27)
Anilines have been shown to be especially susceptible to single-electron oxidation by excited triplet-state photosensitizers (3sens*), and thus, are good potential candidates to probe the oxidative properties of triplet-state chromophoric dissolved organic matter (3CDOM*). However, steady-state experiments tend to underestimate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service