Skip to Content
Merck
All Photos(2)

Key Documents

251186

Sigma-Aldrich

1,3-Bis(trifluoromethyl)benzene

99%

Synonym(s):

α,α,α,α′,α′,α′-Hexafluoro-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CF3)2
CAS Number:
Molecular Weight:
214.11
Beilstein:
2052589
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.379 (lit.)

bp

116-116.3 °C (lit.)

density

1.378 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)c1cccc(c1)C(F)(F)F

InChI

1S/C8H4F6/c9-7(10,11)5-2-1-3-6(4-5)8(12,13)14/h1-4H

InChI key

SJBBXFLOLUTGCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithiation reaction of 1,3-bis(trifluoromethyl)benzene has been investigated[1]. 1,3-Bis(trifluoromethyl)benzene undergoes regioselective metalation and subsequent carboxylation at position 2 to give 2,6-bis(trifluoromethyl)benzoic acid[2].

Application


  • Electrogenerated chemiluminescence of 9,10-diphenylanthracene, rubrene, and anthracene in fluorinated aromatic solvents.: This study explores the electrogenerated chemiluminescence (ECL) properties of various aromatic compounds in fluorinated solvents, including 1,3-Bis(trifluoromethyl)benzene. The research highlights the efficiency and stability of ECL in these fluorinated environments, which can be crucial for applications in high-performance liquid chromatography (HPLC) and other analytical techniques (Vinyard et al., 2008).

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A site selective functionalisation of 1, 3-bis (trifluoromethyl) benzene.
Dmowski W and Piasecka-Maciejewska K.
Tetrahedron, 54(24), 6781-6792 (1998)
The Metalation of 1, 3-Bis (trifluoromethyl) benzene by n butyllithium.
Aeberli P and Houlihan WJ.
Journal of Organometallic Chemistry, 67(3), 321-325 (1974)
Chenyu Lin et al.
BMC chemistry, 14(1), 66-66 (2020-12-10)
For analysis of weak π-π complexes proton-nuclear magnetic resonance (proton-NMR) simultaneously provides information of stacking configurations and association constants [Formula: see text] However, an apparent issue for this approach is inconsistent/impossible constant estimation which often leads to unreasonable interpretation for
Jia Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(26), 14769-14778 (2020-06-17)
Electrophysiological mapping of chronic atrial fibrillation (AF) at high throughput and high resolution is critical for understanding its underlying mechanism and guiding definitive treatment such as cardiac ablation, but current electrophysiological tools are limited by either low spatial resolution or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service