Skip to Content
Merck
All Photos(2)

Key Documents

A9563

Sigma-Aldrich

4-Aminophenylmercuric acetate

≥90% (titration)

Synonym(s):

4-(Acetoxymercurio)aniline, APMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2HgC6H4NH2
CAS Number:
Molecular Weight:
351.75
Beilstein:
3664749
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥90% (titration)

form

powder

mp

163-165 °C (lit.)

solubility

DMSO: soluble 10 mg/mL
H2O: soluble 5 mM
glacial acetic acid: soluble 50 mg/mL

SMILES string

CC(=O)O[Hg]c1ccc(N)cc1

InChI

1S/C6H6N.C2H4O2.Hg/c7-6-4-2-1-3-5-6;1-2(3)4;/h2-5H,7H2;1H3,(H,3,4);/q;;+1/p-1

InChI key

RXSUFCOOZSGWSW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

An organomercurial activator of matrix metalloproteinases.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Bianchi et al.
Osteoarthritis and cartilage, 24(11), 1961-1969 (2016-10-21)
Fibroblast Growth Factor 23 (FGF23) may represent an attractive candidate that could participate to the osteoarthritic (OA)-induced phenotype switch of chondrocytes. To address this hypothesis, we investigated the expression of FGF23, its receptors (FGFRs) and co-receptor (Klotho) in human cartilage
Reihane Ziadlou et al.
Biomolecules, 10(6) (2020-06-25)
In osteoarthritis (OA), inhibition of excessively expressed pro-inflammatory cytokines in the OA joint and increasing the anabolism for cartilage regeneration are necessary. In this ex-vivo study, we used an inflammatory model of human OA chondrocytes microtissues, consisting of treatment with
Jan-Olof Winberg et al.
European journal of biochemistry, 270(19), 3996-4007 (2003-09-27)
In the leukemic macrophage cell-line THP-1, a fraction of the secreted matrix metalloproteinase 9 (MMP-9) is linked to the core protein of chondroitin sulfate proteoglycans (CSPG). Unlike the monomeric and homodimeric forms of MMP-9, the addition of exogenous CaCl2 to
K R French et al.
Equine veterinary journal, 36(3), 261-266 (2004-05-19)
Acute laminitis is characterised by hoof lamellar dermal-epidermal separation at the basement membrane (BM) zone. Hoof lamellar explants cultured in vitro can also be made to separate at the basement membrane zone and investigating how this occurs may give insight
Georgina S Butler et al.
The Journal of biological chemistry, 279(15), 15615-15620 (2004-01-21)
Matrix metalloproteinases (MMPs) are an important family of extracellular proteases that process a variety of biologically significant molecules. MMPs are members of the metzincin superfamily of >770 zinc endopeptidases, which includes astacins, serralysins, adamalysins, leishmanolysins, and snapalysins. Metzincins are characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service