
p-Anisidine

Synonym(s):
4-Aminoanisole, 4-Methoxyaniline, p-Anisidine
Linear Formula:
4-(CH3O)C6H4NH2
CAS Number:
Molecular Weight:
123.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing