A79205
5-Aminoquinoline
97%
Synonym(s):
5-Quinolinamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H8N2
CAS Number:
Molecular Weight:
144.17
Beilstein:
114479
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
powder
bp
310 °C (lit.)
mp
106-109 °C (lit.)
SMILES string
Nc1cccc2ncccc12
InChI
1S/C9H8N2/c10-8-4-1-5-9-7(8)3-2-6-11-9/h1-6H,10H2
InChI key
XMIAFAKRAAMSGX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jagat P Bridhkoti et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(3), 412-417 (2011-04-20)
Photophysical properties of 5-aminoquinoline (5AQ) have been investigated in various non-polar and polar (protic and aprotic) solvents using steady state and time resolved fluorescence. In aprotic solvents, the spectral maxima depend on the polarity. However, in protic solvents both the
V Arjunan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(5), 1215-1223 (2009-10-27)
The Fourier transform infrared (FTIR) and FT-Raman spectra of 5-aminoquinoline (5AQ) have been recorded in the range 4000-400 and 3500-100 cm(-1), respectively. The complete vibrational assignment and analysis of the fundamental modes of the compounds were carried out using the
R Banoo et al.
Indian journal of biochemistry & biophysics, 30(4), 229-233 (1993-08-01)
In vitro oxidation and acetylation of 5-aminoquinoline by rabbit liver enzyme preparation has been investigated. Incubation of 5-aminoquinoline with cytosol fraction of the enzyme preparation in the presence of acetyl coenzyme-A gave rise to three different products, viz. 5-amino-2-hydroxy quinoline
B G Siim et al.
Biochemical pharmacology, 48(8), 1593-1604 (1994-10-18)
The 4-alkylamino-5-nitroquinolines (5NQs) are a new series of bioreductive drugs that exhibit varying degrees of selective toxicity (up to 60-fold) under hypoxic conditions in cell culture. This study tested the hypothesis that differences in hypoxia-selective cytotoxicity in this series reflect
Mark A Eckert et al.
Nature, 569(7758), 723-728 (2019-05-03)
High-grade serous carcinoma has a poor prognosis, owing primarily to its early dissemination throughout the abdominal cavity. Genomic and proteomic approaches have provided snapshots of the proteogenomics of ovarian cancer1,2, but a systematic examination of both the tumour and stromal
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service