A41402
4-Aminobenzophenone
Synonym(s):
4-Aminophenyl phenyl ketone, 4-Benzoylaniline
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
H2NC6H4C(O)C6H5
CAS Number:
Molecular Weight:
197.23
Beilstein:
389292
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
powder
mp
121-124 °C (lit.)
SMILES string
Nc1ccc(cc1)C(=O)c2ccccc2
InChI
1S/C13H11NO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9H,14H2
InChI key
RBKHNGHPZZZJCI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ghassan Abu Sheikha et al.
Chemical & pharmaceutical bulletin, 66(4), 423-426 (2018-04-03)
Hyperlipidemia is known as an elevation of plasma lipid components. It contributes significantly to atherosclerosis which is one of the most important causative factors in cardiovascular diseases. Agents that cause a dramatic decrease in serum lipid levels are of great
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service