Skip to Content
Merck
All Photos(1)

Key Documents

96324

Sigma-Aldrich

1-Methyl-3-octylimidazolium tetrafluoroborate

≥97.0% (HPLC)

Synonym(s):

OMIMBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23BF4N2
CAS Number:
Molecular Weight:
282.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

electrochemical window 6.0 V

Quality Level

Assay

≥97.0% (HPLC)

form

liquid

conductivity

0.43 mS/cm

viscosity

422 stokes

density

1.12 g/mL at 20 °C (lit.)

SMILES string

F[B-](F)(F)F.CCCCCCCC[n+]1ccn(C)c1

InChI

1S/C12H23N2.BF4/c1-3-4-5-6-7-8-9-14-11-10-13(2)12-14;2-1(3,4)5/h10-12H,3-9H2,1-2H3;/q+1;-1

InChI key

GXZCAMSPWNHTAE-UHFFFAOYSA-N

Other Notes

Enzyme catalysis in ionic liquids: lipase-catalyzed kinetic resolution; phase behaviour

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.H. Schofer et al.
Chemical Communications (Cambridge, England), 425-425 (2001)
J.D. Holbrey et al.
J. Chem. Soc., Dalton Trans., 2133-2133 (1999)
Vasiliki Tsarpali et al.
Aquatic toxicology (Amsterdam, Netherlands), 203, 88-94 (2018-08-14)
The current study is based on the increasing demand for the assessment of ionic liquid (IL)-mediated aquatic toxicity. Specifically, although a lot of studies have been performed so far, investigating IL-mediated adverse effects on numerous aquatic organisms, little is known
Sanjay V Malhotra et al.
Bioorganic & medicinal chemistry letters, 20(2), 581-585 (2009-12-17)
The anti-cancer activity and cytotoxicity of imidazolium-based ionic liquids has been determined for the first time via NCI's in vitro 60 human tumor cell lines. The preliminary SAR showed that the chain length of alkyl substitution at N-3 position of
Saheli Mitra et al.
Biochimica et biophysica acta. Biomembranes, 1863(6), 183589-183589 (2021-03-03)
Ionic liquids (ILs) are the attractions of researchers today due to their vast area of potential applications. For biomedical uses, it becomes essential to understand their interactions with cellular membrane. Here, the membrane is mimicked with lipid bilayer and monolayer

Articles

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service